6-(Morpholin-4-yl)pyridine-2-carbonitrile 98%

Lieferant: Apollo Scientific

OR959809-1G
APOSOR959809-1GEA 348 CHF
APOSOR959809-1G
6-(Morpholin-4-yl)pyridine-2-carbonitrile 98%
6-(Morpholin-4-yl)pyridine-2-carbonitrile
Formel: C₁₀H₁₁N₃O
MW: 189,214 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD08690244
CAS-Nummer: 868755-53-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR