2-Cyclopropylaminonicotinic acid 95%

Lieferant: Apollo Scientific

Synonyms: 2-(Cyclopropylamino)nicotinic acid

OR958277-10G OR958277-5G
APOSOR958277-10GEA 69.6 CHF
APOSOR958277-10G APOSOR958277-5G
2-Cyclopropylaminonicotinic acid 95%
2-Cyclopropylaminonicotinic acid
Formel: C₉H₁₀N₂O₂
MW: 178.188 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD11106728
CAS-Nummer: 639807-18-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR