2-Ethoxy-5-methoxy-phenol 95%

Lieferant: Apollo Scientific

OR957539-1G
APOSOR957539-1GEA 758 CHF
APOSOR957539-1G
2-Ethoxy-5-methoxy-phenol 95%
2-Ethoxy-5-methoxy-phenol
Formel: C₉H₁₂O₃
MW: 168,19 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD24717691
CAS-Nummer: 90534-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR