4-Chloro-N-(2-hydroxyethyl)benzamide 95%

Lieferant: Apollo Scientific

OR957106-1G
APOSOR957106-1GEA 645 CHF
APOSOR957106-1G
4-Chloro-N-(2-hydroxyethyl)benzamide 95%
4-Chloro-N-(2-hydroxyethyl)benzamide
Formel: C₉H₁₀ClNO₂
MW: 199,634 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00461164
CAS-Nummer: 7400-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR