1-Chloro-3-methyl-5-(propan-2-yloxy)benzene 95%

Lieferant: Apollo Scientific

OR957095-1G
APOSOR957095-1GEA 758 CHF
APOSOR957095-1G
1-Chloro-3-methyl-5-(propan-2-yloxy)benzene 95%
1-Chloro-3-methyl-5-(propan-2-yloxy)benzene
Formel: C₁₀H₁₃ClO
MW: 184,663 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD23113809
CAS-Nummer: 1369862-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR