Cyclopropyl methoxylamine hydrochloride 95%

Lieferant: Apollo Scientific

Synonyms: O-(Cyclopropylmethyl)hydroxylamine hydrochloride

OR953045-1G
APOSOR953045-1GEA 166 CHF
APOSOR953045-1G
Cyclopropyl methoxylamine hydrochloride 95%
Cyclopropyl methoxylamine hydrochloride
Formel: C₄H₉NO·ClH
MW: 123,581 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD08689925
CAS-Nummer: 74124-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR