3-Amino-1-adamantanol

Lieferant: Apollo Scientific
Achtung

Synonyms: 3-Aminoadamantan-1-ol

OR9509-100G OR9509-25G OR9509-5G OR9509-500G
APOSOR9509-100GEA 29.7 CHF
APOSOR9509-100G APOSOR9509-25G APOSOR9509-5G APOSOR9509-500G
3-Amino-1-adamantanol
3-Amino-1-adamantanol
Formel: C₁₀H₁₇NO
MW: 167.25 g/mol
Melting Pt: 263…269 °C (Zers.)
Flash-Pt: 110 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD01821204
CAS-Nummer: 702-82-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR