5-Chloro-2-(methylamino)phenol 95%

Lieferant: Apollo Scientific

OR942472-1G OR942472-250MG
APOSOR942472-1GEA 688 CHF
APOSOR942472-1G APOSOR942472-250MG
5-Chloro-2-(methylamino)phenol 95%
5-Chloro-2-(methylamino)phenol
Formel: C₇H₈ClNO
MW: 157.598 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD16998610
CAS-Nummer: 1243357-05-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR