2-Chloro-4-methoxybenzothiazole 95%

Lieferant: Apollo Scientific

Synonyms: 2-Chloro-4-methoxybenzo[d]thiazole

OR939266-1G
APOSOR939266-1GEA 278 CHF
APOSOR939266-1G
2-Chloro-4-methoxybenzothiazole 95%
2-Chloro-4-methoxybenzothiazole
Formel: C₈H₆ClNOS
MW: 199,657 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD04448819
CAS-Nummer: 3507-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR