Methyl-3-isocyanatopropanoate 95%

Lieferant: Apollo Scientific

Synonyms: Methyl 3-isocyanatopropanoate

OR938571-1G
APOSOR938571-1GEA 226 CHF
APOSOR938571-1G
Methyl-3-isocyanatopropanoate 95%
Methyl-3-isocyanatopropanoate

  • May form small amount of sediment.

Formel: C₅H₇NO₃
MW: 129.12 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00797846
CAS-Nummer: 50835-77-3

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR