Pyridazin-4-ylmethanamine dihydrochloride

Lieferant: Apollo Scientific

OR933480-1G
APOSOR933480-1GEA 974 CHF
APOSOR933480-1G
Pyridazin-4-ylmethanamine dihydrochloride
Pyridazin-4-ylmethanamine dihydrochloride
Formel: C₅H₇N₃·2ClH
MW: 182.051 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD27756609
CAS-Nummer: 1028615-75-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR