(1H-Pyrrol-2-yl)methanamine hydrochloride

Lieferant: Apollo Scientific

OR933425-1G OR933425-250MG OR933425-5G
APOSOR933425-1GEA 140 CHF
APOSOR933425-1G APOSOR933425-250MG APOSOR933425-5G
(1H-Pyrrol-2-yl)methanamine hydrochloride
(1H-Pyrrol-2-yl)methanamine hydrochloride
Formel: C₅H₉ClN₂
MW: 132.59 g/mol
Melting Pt: 120…140 °C
Lager Temperatur: Kühlschrank
MDL Number: MFCD20037390
CAS-Nummer: 1351479-09-8

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR