2-Ethylpyrimidine-5-carboxylic acid 95%

Lieferant: Apollo Scientific

OR930977-1G
APOSOR930977-1GEA 522 CHF
APOSOR930977-1G
2-Ethylpyrimidine-5-carboxylic acid 95%
2-Ethylpyrimidine-5-carboxylic acid
Formel: C₇H₈N₂O₂
MW: 152,151 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD09054863
CAS-Nummer: 72790-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR