2-[(2-Methylphenyl)methyl]pyrrolidine 95%

Lieferant: Apollo Scientific

OR929958-1G
APOSOR929958-1GEA 602 CHF
APOSOR929958-1G
2-[(2-Methylphenyl)methyl]pyrrolidine 95%
2-[(2-Methylphenyl)methyl]pyrrolidine
Formel: C₁₂H₁₇N
MW: 175,27 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD02663439
CAS-Nummer: 383127-27-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR