(3-Propoxyphenyl)methanamine hydrochloride 95%

Lieferant: Apollo Scientific

Synonyms: (3-Propoxyphenyl)methanamine

OR927946-1G OR927946-250MG
APOSOR927946-1GEA 497 CHF
APOSOR927946-1G APOSOR927946-250MG
(3-Propoxyphenyl)methanamine hydrochloride 95%
(3-Propoxyphenyl)methanamine hydrochloride
Formel: C₁₀H₁₆ClNO
MW: 201.7 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD10687270
CAS-Nummer: 37806-33-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR