1-Methyl-7-azaindole-4-carboxylic acid 95%

Lieferant: Apollo Scientific

OR927503-1G OR927503-250MG
APOSOR927503-1GEA 154 CHF
APOSOR927503-1G APOSOR927503-250MG
1-Methyl-7-azaindole-4-carboxylic acid 95%
1-Methyl-7-azaindole-4-carboxylic acid
Formel: C₉H₈N₂O₂
Lager Temperatur: Raumtemperatur
MDL Number: MFCD13302392
CAS-Nummer: 1147753-38-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR