Ethyl-3-isopropyl pyrazole-4-carboxylate 95%

Lieferant: Apollo Scientific

Synonyms: Ethyl 3-isopropyl-1H-pyrazole-4-carboxylate

OR924858-1G OR924858-250MG
APOSOR924858-1GEA 584 CHF
APOSOR924858-1G APOSOR924858-250MG
Ethyl-3-isopropyl pyrazole-4-carboxylate 95%
Ethyl-3-isopropyl pyrazole-4-carboxylate
Formel: C₉H₁₄N₂O₂
MW: 182.22 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD07369096
CAS-Nummer: 342026-17-9

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR