(3-Chloropropyl)sulphamoyl chloride 95%

Lieferant: Apollo Scientific

OR923366-1G
APOSOR923366-1GEA 645 CHF
APOSOR923366-1G
(3-Chloropropyl)sulphamoyl chloride 95%
(3-Chloropropyl)sulphamoyl chloride
Formel: C₃H₇Cl₂NO₂S
MW: 192,064 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD14529128
CAS-Nummer: 42065-72-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR