sec-Butyl ethyl ether 95%

Lieferant: Apollo Scientific

OR922803-25G OR922803-5G
APOSOR922803-25GEA 0 CHF
APOSOR922803-25G APOSOR922803-5G
sec-Butyl ethyl ether 95%
sec-Butyl ethyl ether
Formel: C₆H₁₄O
MW: 102.18 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00053773
CAS-Nummer: 2679-87-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR