Methyl 3-methyl-1H-pyrrole-2-carboxylate 95%

Lieferant: Apollo Scientific

OR921497-1G
APOSOR921497-1GEA 152 CHF
APOSOR921497-1G
Methyl 3-methyl-1H-pyrrole-2-carboxylate 95%
Methyl 3-methyl-1H-pyrrole-2-carboxylate
Formel: C₇H₉NO₂
Lager Temperatur: Raumtemperatur
MDL Number: MFCD09754196
CAS-Nummer: 40611-69-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR