1-Brom-3-methylbutan

Lieferant: Apollo Scientific

Synonyms: 3-Methylbutylbromide

OR921443-500G
APOSOR921443-500GEA 634 CHF
APOSOR921443-500G
1-Brom-3-methylbutan
1-Brom-3-methylbutan
Formel: C₅H₁₁Br
MW: 151.05 g/mol
Siedepunkt: 120…121 °C
Melting Pt: –112 °C
Dichte: 1.261 g/cm³ (25 °C)
Flash-Pt: 22 °C
Lager Temperatur: Kühlschrank
MDL Number: MFCD00000253
CAS-Nummer: 107-82-4
UN: 3241
ADR: 4.1 III
Merck Index: 13,05130

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR