N, N'-Diacetyl-1,4-Phenylendiamin 95%

Lieferant: Apollo Scientific

Synonyms: DAPPD, N,N'-1,4-Phenylenebisacetamide, N,N'-(1,4-Phenylene)diacetamide

OR920370-25G
APOSOR920370-25GEA 207 CHF
APOSOR920370-25G
N, N'-Diacetyl-1,4-Phenylendiamin 95%
N, N'-Diacetyl-1,4-Phenylendiamin
Formel: C₁₀H₁₂N₂O₂
MW: 192.22 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00026142
CAS-Nummer: 140-50-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR