ɑ,ɑ-Dimethyl-γ-butyrolactone 95%

Lieferant: Apollo Scientific

Synonyms: Dihydro-3,3-dimethyl-2(3H)-furanone, α,α-Dimethyl-γ-butyrolactone

OR920353-1G
APOSOR920353-1GEA 436 CHF
APOSOR920353-1G
ɑ,ɑ-Dimethyl-γ-butyrolactone 95%
alpha,alpha-Dimethyl-gamma-butyrolactone
Formel: C₆H₁₀O₂
MW: 114.14 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD03844788
CAS-Nummer: 3709-08-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR