Methyl-4-acetoxybenzoat  95%

Lieferant: Apollo Scientific

Synonyms: 4-Acetoxybenzoic Acid Methyl Ester, Methyl 4-acetoxybenzoate

OR919737-25G
APOSOR919737-25GEA 105 CHF
APOSOR919737-25G
Methyl-4-acetoxybenzoat  95%
Methyl-4-acetoxybenzoat 
Formel: C₁₀H₁₀O₄
MW: 194,19 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00017517
CAS-Nummer: 24262-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR