Disodium-4-formylbenzene-1,3-disulphonate 98+%

Lieferant: Apollo Scientific
Achtung

Synonyms: 4-Formylbenzene-1,3-disulphonic acid disodium salt, Sodium 4-formylbenzene-1,3-disulphonate

OR919217-500G
APOSOR919217-500GEA 752 CHF
APOSOR919217-500G
Disodium-4-formylbenzene-1,3-disulphonate 98+%
Disodium-4-formylbenzene-1,3-disulphonate
Formel: C₇H₆O₇S₂
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00043913
CAS-Nummer: 33513-44-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR