N-tert-Butyl-2-nitroaniline 98%

Lieferant: Apollo Scientific

OR913071-1G OR913071-25G OR913071-5G
APOSOR913071-1GEA 174 CHF
APOSOR913071-1G APOSOR913071-25G APOSOR913071-5G
N-tert-Butyl-2-nitroaniline 98%
N-tert-Butyl-2-nitroaniline
Formel: C₁₀H₁₄N₂O₂
MW: 194.23 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD16170799
CAS-Nummer: 28458-45-9

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR