N,N'-Di-sec-butylcarbodiimide

Lieferant: Apollo Scientific

OR9106-1G
APOSOR9106-1GEA 184 CHF
APOSOR9106-1G
N,N'-Di-sec-butylcarbodiimide
N,N'-Di-sec-butylcarbodiimide
Formel: C₉H₁₈N₂
Lager Temperatur: Raumtemperatur
MDL Number: MFCD01861858
CAS-Nummer: 66006-67-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR