Methyl-4-pentenoat 95%

Lieferant: Apollo Scientific

Synonyms: Methyl pent-4-enoate

OR908012-5G
APOSOR908012-5GEA 90.5 CHF
APOSOR908012-5G
Methyl-4-pentenoat 95%
Methyl-4-pentenoat
Formel: C₆H₁₀O₂
MW: 114.14 g/mol
Dichte: 0.922 g/cm³ (25 °C)
Flash-Pt: 29 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD03990590
CAS-Nummer: 818-57-5
UN: 3272
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR