Tetrahydropyran-2-carbonsäure 97%

Lieferant: Apollo Scientific

Synonyms: Tetrahydro-2H-pyran-2-carboxylic acid

OR907468-1G
APOSOR907468-1GEA 62.7 CHF
APOSOR907468-1G
Tetrahydropyran-2-carbonsäure 97%
Tetrahydropyran-2-carbonsäure
Formel: C₆H₁₀O₃
MW: 130.1418 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD07779239
CAS-Nummer: 51673-83-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR