Pyrazinamid 98%

Lieferant: Apollo Scientific
Achtung

Synonyms: Pyrazinecarboxamide, Pyrazine-2-carboxamide, 2-Pyrazinecarboxamide

OR9043-100G OR9043-500G
APOSOR9043-100GEA 155 CHF
APOSOR9043-100G APOSOR9043-500G
Pyrazinamid 98%
Pyrazinamid
Formel: C₅H₅N₃O
Melting Pt: 189…191 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00006132
CAS-Nummer: 98-96-4
EINECS: 202-717-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR