3-(3-Chlorophenyl)propan-1-ol 96%

Lieferant: Apollo Scientific

OR900159-1G
APOSOR900159-1GEA 323 CHF
APOSOR900159-1G
3-(3-Chlorophenyl)propan-1-ol 96%
3-(3-Chlorophenyl)propan-1-ol
Formel: C₉H₁₁ClO
MW: 170.636 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD09028713
CAS-Nummer: 22991-03-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR