4'-Chlor-2'-hydroxyacetophenon

Lieferant: Apollo Scientific
Achtung

Synonyms: 1-(4-Chloro-2-hydroxyphenyl)ethan-1-one, 4-Chloro-2-hydroxyacetophenone

OR8803-10G OR8803-50G
APOSOR8803-10GEA 155 CHF
APOSOR8803-10G APOSOR8803-50G
4'-Chlor-2'-hydroxyacetophenon
4'-Chlor-2'-hydroxyacetophenon
Formel: C₈H₇ClO₂
MW: 170.6
Siedepunkt: 247 °C
Melting Pt: 27…30 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00238557
CAS-Nummer: 6921-66-0

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR