1-(Bromomethyl)pyrene

Lieferant: Apollo Scientific
Gefahr

Synonyms: 3-(Bromomethyl)pyrene

OR8551T-500MG
APOSOR8551T-500MGEA 401 CHF
APOSOR8551T-500MG
1-(Bromomethyl)pyrene
1-(Bromomethyl)pyrene
Formel: C₁₇H₁₁Br
Melting Pt: 140…145 °C
Lager Temperatur: Gefrierschrank
MDL Number: MFCD00269937
CAS-Nummer: 2595-90-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR