Thieno[3,2-b]thiophene-2-carbohydrazide

Lieferant: Apollo Scientific
Achtung

OR8551-1G
APOSOR8551-1GEA 307 CHF
APOSOR8551-1G
Thieno[3,2-b]thiophene-2-carbohydrazide
Thieno[3,2-b]thiophene-2-carbohydrazide
Formel: C₇H₆N₂OS₂
MW: 198.27 g/mol
Melting Pt: 188…190 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD04112097
CAS-Nummer: 685114-87-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR