3-Amino-4-brom-1-methyl-1H-pyrazol 98%

Lieferant: Apollo Scientific
Achtung

Synonyms: 4-Bromo-1-methyl-1H-pyrazol-3-amine, 3-Amino-4-bromo-1-methyl-1H-pyrazole, 3-Amino-4-bromo-1-methylpyrazole

OR7682-5G
APOSOR7682-5GEA 90.5 CHF
APOSOR7682-5G
3-Amino-4-brom-1-methyl-1H-pyrazol 98%
3-Amino-4-bromo-1-methyl-1H-pyrazole
Formel: C₄H₆BrN₃
MW: 176.02 g/mol
Melting Pt: 96…100 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00663065
CAS-Nummer: 146941-72-2

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR