4-(Aminomethyl)phenol

Lieferant: Apollo Scientific
Gefahr

Synonyms: 4-Hydroxybenzylamine

OR7611-5G OR7611-25G OR7611-100G
APOSOR7611-5GEA 26.2 CHF
APOSOR7611-5G APOSOR7611-25G APOSOR7611-100G
4-(Aminomethyl)phenol
4-(Aminomethyl)phenol
Formel: C₇H₉NO
MW: 123.16 g/mol
Melting Pt: 116…118 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD19690971
CAS-Nummer: 696-60-6
UN: 3259
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR