4-Nonylphenylboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyms: 4-N-Nonylphenylboronic acid, 4-n-Nonylbenzeneboronic acid, FAAH/MAGL-IN-4

OR7244-1G OR7244-5G
APOSOR7244-1GEA 190 CHF
APOSOR7244-1G APOSOR7244-5G
4-Nonylphenylboronsäure
4-Nonylphenylboronsäure
Formel: C₁₅H₂₅BO₂
Melting Pt: 82…85 °C
Lager Temperatur: Kühlschrank
MDL Number: MFCD02093070
CAS-Nummer: 256383-45-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR