4,4,5,5-Tetramethyl-1,3,2-dioxaborolan

Lieferant: Apollo Scientific
Gefahr

Synonyms: Pinacol Borane

OR7123-100G OR7123-25G OR7123-5G
APOSOR7123-100GEA 174 CHF
APOSOR7123-100G APOSOR7123-25G APOSOR7123-5G
4,4,5,5-Tetramethyl-1,3,2-dioxaborolan
4,4,5,5-Tetramethyl-1,3,2-dioxaborolan

Reportedly couples with aryl halides to produce aryl boronates:J.Org.Chem.(1997)62,6458-6459.

Formel: C₆H₁₃BO₂
Siedepunkt: 42…43 °C/50mmHg
Dichte: 0.882 g/cm³ (25 °C)
Flash-Pt: 5 °C
Lager Temperatur: Kühlschrank
MDL Number: MFCD00674030
CAS-Nummer: 25015-63-8
UN: 3399
ADR: 4.3+3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR