Methyl 3-methyl-1H-indole-5-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyms: 5-(Methoxycarbonyl)-3-methyl-1H-indole

OR62081-1G OR62081-250MG
APOSOR62081-1GEA 619 CHF
APOSOR62081-1G APOSOR62081-250MG
Methyl 3-methyl-1H-indole-5-carboxylate
Methyl 3-methyl-1H-indole-5-carboxylate
Formel: C₁₁H₁₁NO₂
Lager Temperatur: Kühlschrank
MDL Number: MFCD11975645
CAS-Nummer: 588688-33-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR