Pinakol

Lieferant: Apollo Scientific
Gefahr

Synonyms: 2,3-Dimethylbutane-2,3-diol, Tetramethylethylene glycol, 2,3-Dimethyl-2,3-butanediol

OR61505-100G OR61505-25G OR61505-500G
APOSOR61505-100GEA 41.8 CHF
APOSOR61505-100G APOSOR61505-25G APOSOR61505-500G
Pinakol
Pinakol
Formel: C₆H₁₄O₂
Siedepunkt: 174 °C
Melting Pt: 40…43 °C
Flash-Pt: 77 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00004462
CAS-Nummer: 76-09-5
EINECS: 200-933-5
UN: 1325
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR