Diethyl iminodiacetate 98%

Lieferant: Apollo Scientific
Achtung

Synonyms: Iminodiacetic Acid Diethyl Ester, Diethyl 2,2'-azanediyldiacetate

OR61269-100G
APOSOR61269-100GEA 50.6 CHF
APOSOR61269-100G
Diethyl iminodiacetate 98%
Diethyl iminodiacetate
Formel: C₈H₁₅NO₄
MW: 189.21 g/mol
Siedepunkt: 208 °C
Dichte: 1.056 g/cm³ (25 °C)
Flash-Pt: 113 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00041925
CAS-Nummer: 6290-05-7
EINECS: 228-533-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR