4-(4-Chlorbenzoyl)pyridin

Lieferant: Apollo Scientific
Achtung

OR59834-1G OR59834-5G
APOSOR59834-1GEA 119 CHF
APOSOR59834-1G APOSOR59834-5G
4-(4-Chlorbenzoyl)pyridin
4-(4-Chlorbenzoyl)pyridin
Formel: C₁₂H₈ClNO
MW: 217.66 g/mol
Melting Pt: 107…109 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00006431
CAS-Nummer: 14548-48-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR