8-Hydroxyquinoline sulphate (2:1) monopotassium salt

Lieferant: Apollo Scientific
Achtung

Synonyms: Potassium hydrogen sulphate - quinolin-8-ol (1:2), 8-Hydroxyquinoline potassium sulphate

OR59477-25G
APOSOR59477-25GEA 105 CHF
APOSOR59477-25G
8-Hydroxyquinoline sulphate (2:1) monopotassium salt
8-Hydroxyquinoline sulphate (2:1) monopotassium salt
Formel: C₉H₇NO·K·HO₄S
Lager Temperatur: Raumtemperatur
MDL Number: MFCD08062407
CAS-Nummer: 15077-57-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR