1,5-Dimethyl-1H-indole-2-carbaldehyde 95+%

Lieferant: Apollo Scientific

OR5667-250MG
APOSOR5667-250MGEA 152 CHF
APOSOR5667-250MG
1,5-Dimethyl-1H-indole-2-carbaldehyde 95+%
1,5-Dimethyl-1H-indole-2-carbaldehyde
Formel: C₁₁H₁₁NO
MW: 173,22 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD06800611
CAS-Nummer: 883526-76-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR