1-Benzhydryl-2-thiourea

Lieferant: Apollo Scientific
Achtung

OR5642-250MG
APOSOR5642-250MGEA 69.6 CHF
APOSOR5642-250MG
1-Benzhydryl-2-thiourea
1-Benzhydryl-2-thiourea
Formel: C₁₄H₁₄N₂S
MW: 242.35 g/mol
Melting Pt: 184 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00060461
CAS-Nummer: 92192-94-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR