2-Brom-5-thiazolcarbonsäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyms: 2-Bromothiazole-5-carboxylic acid

OR5591-25G
APOSOR5591-25GEA 119 CHF
APOSOR5591-25G
2-Brom-5-thiazolcarbonsäure 97%
2-Bromo-5-thiazolecarboxylic acid
Formel: C₄H₂BrNO₂S
MW: 208,04 g/mol
Melting Pt: 180…184 °C
Lager Temperatur: Kühlschrank
MDL Number: MFCD04115730
CAS-Nummer: 54045-76-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR