2-Aminopyrazin

Lieferant: Apollo Scientific
Achtung

Synonyms: Aminopyrazine, Pyrazin-2-amine

OR5470-100G OR5470-25G OR5470-500G
APOSOR5470-100GEA 57.5 CHF
APOSOR5470-100G APOSOR5470-25G APOSOR5470-500G
2-Aminopyrazin
2-Aminopyrazine
Formel: C₄H₅N₃
Melting Pt: 118…121 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00006137
CAS-Nummer: 5049-61-6
EINECS: 225-748-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR