Fmoc-O-trityl-L-threonine 98.5+%

Lieferant: Apollo Scientific
Achtung

Synonyms: Fmoc-Thr(Trt)-OH, (2S,3R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(trityloxy)butanoic acid

OR53138-1G OR53138-25G OR53138-5G
APOSOR53138-1GEA 158 CHF
APOSOR53138-1G APOSOR53138-25G APOSOR53138-5G
Fmoc-O-trityl-L-threonine 98.5+%
Fmoc-O-trityl-L-threonine
Formel: C₃₈H₃₃NO₅
MW: 583.684 g/mol
Melting Pt: 70…75 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00237034
CAS-Nummer: 133180-01-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR