6-Bromo-2-chloro-7-methyl-3H-imidazo[4,5-b]pyridine

Lieferant: Apollo Scientific
Achtung

Synonyms: 5-Bromo-2-chloro-4-methyl-7-azabenzimidazole

OR52096-1G OR52096-250MG
APOSOR52096-1GEA 1200 CHF
APOSOR52096-1G APOSOR52096-250MG
6-Bromo-2-chloro-7-methyl-3H-imidazo[4,5-b]pyridine
6-Bromo-2-chloro-7-methyl-3H-imidazo[4,5-b]pyridine
Formel: C₇H₅BrClN₃
MW: 246.492 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD22460484
CAS-Nummer: 1388071-17-7

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR