N,O-Bis(trimethylsilyl)acetamid 97%

Lieferant: Apollo Scientific
Gefahr

Synonyms: BSA, Silyl Derivatisation, Trimethylsilyl N-(trimethylsilyl)acetimidate, Trimethylsilyl N-trimethylsilylacetamidate

OR5155A-100G OR5155A-500G OR5155A-25G
APOSOR5155A-100GEA 34.8 CHF
APOSOR5155A-100G APOSOR5155A-500G APOSOR5155A-25G
N,O-Bis(trimethylsilyl)acetamid 97%
N,O-Bis(trimethylsilyl)acetamid
Formel: C₈H₂₁NOSi₂
Siedepunkt: 71…73 °C/35mmHg
Melting Pt: 24 °C
Dichte: 0.832 g/cm³ (25 °C)
Flash-Pt: 42 °C
Lager Temperatur: Kühlschrank
MDL Number: MFCD00008270
CAS-Nummer: 10416-59-8
EINECS: 233-892-7
UN: 2920
ADR: 8+3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR